Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KLF2 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$106.19 - $320.71

Specifications

Antigen KLF2
Dilution Western Blot 1.0 ug/ml, Chromatin Immunoprecipitation, Immunohistochemistry
Applications Western Blot, ChIP assay, Immunohistochemistry
Conjugate Unconjugated
Format Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB126811
View Documents
Novus Biologicals
NBP310956100UL
100 μg
Each for $320.71
Add to cart
 
NB126812
View Documents
Novus Biologicals
NBP31095625UL
25 μg
Each for $106.19
Add to cart
 
Description

Description

KLF2 Polyclonal antibody specifically detects KLF2 in Human samples. It is validated for Western Blot, ChIP assay, Immunohistochemistry
Specifications

Specifications

KLF2
Western Blot, ChIP assay, Immunohistochemistry
Purified
RUO
PBS buffer, 2% sucrose
10365
Polyclonal
Affinity purified
Western Blot 1.0 ug/ml, Chromatin Immunoprecipitation, Immunohistochemistry
Unconjugated
Rabbit
Human
Krueppel-like factor 2, Kruppel-like factor 2 (lung), Kruppel-like factor LKLF, LKLFLung krueppel-like factor, lung Kruppel-like zinc finger transcription factor
The immunogen is a synthetic peptide directed towards the middle region of human KLF2 (NP_057354). Peptide sequence PPAFGLFDDAAAAAAALGLAPPAARGLLTPPASPLELLEAKPKRGRRSWP
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit