Learn More
KLF4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier:  Thermo Scientific PA1095
Catalog No. PIPA1095
Add to Cart
Description
Description
The predicted molecular weight of KLF4 is ∽55kD. The migration of this protein by SDS-PAGE was observed slightly higher (∽60kD) with both PA1-095 and a benchmark antibody. PA1-095 detects a predominant band at ∽60kD, and a nonspecific band of unknown origin at ∽80kD in both whole cell lysates and nuclear extracts. For best results, StartingBlock T20 (TBS) Blocking Buffer (Product # 37543) is recommended.
KLF4 Polyclonal antibody specifically detects KLF4 in Human, Mouse samples. It is validated for Immunocytochemistry, Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
KLF4
Polyclonal
Unconjugated
KLF4
endothelial Kruppel-like zinc finger protein; epithelial zinc finger protein EZF; Ezf; Gklf; gut Kruppel-like factor; gut-enriched krueppel-like factor; high mobility group nucleosomal binding domain 3; high mobility group nucleosome-binding domain-containing protein 3; HMGN3; KLF4; Krueppel-like factor 4; Kruppel like factor 4; Kruppel-like factor 4 (Gut enriched kruppel-like factor) (Epithelial zinc-finger protein EZF); Kruppel-like factor 4 (gut); PNAS-24; PNAS-25; RP11-150J11.1; thyroid hormone receptor interacting protein 7; Thyroid receptor-interacting protein 7; TR-interacting protein 7; TRIP7; Zie
Rabbit
Protein A
RUO
16600, 9314
-20°C
Liquid
Immunocytochemistry, Western Blot
0.5 mg/mL
PBS with 1mg/mL BSA and 0.05% sodium azide
O43474, Q60793
KLF4
Recombinant N-terminal fragment (amino acids 1-118) of human KLF4
200 μL
Primary
Human, Mouse
Antibody
IgG
Recommendations
Recommendations