Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KLF5 Monoclonal Antibody (OTI6D2), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene CF811868


Description
KLF5 Monoclonal antibody specifically detects KLF5 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
KLF5 | |
Monoclonal | |
See Label | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
O43736, Q61500 | |
Mouse | |
Protein A/G | |
RUO | |
16431, 317218, 9452 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
OTI6D2 | |
Unconjugated | |
ITM2A | |
(intestinal Kruppel-like factor; 4930520J07Rik; basic transcription element binding protein 2; basic transcription element binding protein BTEB2; basic transcription element-binding protein 2; BTEB2; BTE-binding protein 2; CKLF; colon krueppel-like factor; colon kruppel-like factor; GC box binding protein 2; GC-box-binding protein 2; IKLF; Intestinal-enriched krueppel-like factor; intestinal-enriched kruppel-like factor; Klf5; Klf5C isoform; Krueppel-like factor 5; Kruppel like factor 5; Kruppel-like factor 5; Kruppel-like factor 5 (intestinal); RP11-505F3.5; transcription factor BTEB2 | |
Full length human recombinant protein of human ITM2A (NP_004858) produced in E.coli. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Lyophilized |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction