Learn More
KLF5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 210171AP
Catalog No. 50-173-3206
Add to cart
Description
Description
This antibody detects the 50-60 kDa isoform 1 of KLF5. The antibody can recognize isoform 2 (33 kDa) and isoform 3 (18 kDa) too.
KLF5 is a member of the Kruppel-like factor subfamily of zinc finger proteins. Since the protein localizes to the nucleus and binds the epidermal growth factor response element, the protein is thought to be a transcription factor.
Specifications
Specifications
KLF5
0.4 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q13887, Q9Z0Z7
BTE binding protein 2, BTEB2, CKLF, Colon krueppel like factor, GC box binding protein 2, IKLF, KLF5, Krueppel like factor 5, Transcription factor BTEB2
Rabbit
IgG
150 μL
-20°C
Primary
12224, 688, 84410
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
KLF5
Liquid
Klf5
KLF5 Fusion Protein Ag15267
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat