Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KLF6 SV1 Rabbit anti-Human, Non-human Primate, Polyclonal, FabGennix
Rabbit Polyclonal Antibody
Supplier: Fabgennix Inc KLF6SV1101AP

Description
KLF6 SV1 Polyclonal antibody specifically detects KLF6 SV1 in Human, Non-human Primate samples. It is validated for ELISA, Immunohistochemistry, Immunoprecipitation, Western BlotSpecifications
KLF6 SV1 | |
Polyclonal | |
Unconjugated | |
KLF6 | |
KLF6; Proto-oncogene BCD1; Transcription factor Zf9 | |
Synthetic peptide taken within amino acid region 170-220 on human KLF6 protein. | |
200 μL | |
Primary | |
Human, Non-human Primate | |
Antibody | |
IgG |
ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot | |
0.5-1.5 mg/mL | |
proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8 | |
Q99612 | |
Rabbit | |
Affinity chromatography | |
RUO | |
1316, 450269 | |
-20°C | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction