Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KLF7 Mouse anti-Human, Alexa Fluor™ 532, Clone: 3G3, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP242626AF532

Catalog No. NP242626A32


Add to cart

Description

Description

KLF7 Monoclonal antibody specifically detects KLF7 in Human samples. It is validated for Western Blot,ELISA.
Specifications

Specifications

KLF7
3G3
Western Blot, ELISA
Krueppel-like factor 7, Kruppel-like factor 7 (ubiquitous), ubiquitous Kruppel-like transcription factor, UKLFUbiquitous krueppel-like factor
Mouse
IgG1 Kappa
Store at 4°C in the dark.
Monoclonal
Human
ELISA, Western Blot
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
KLF7
Recombinant human KLF7 (1-302aa) purified from E. coli
0.1 mL
Primary
8609
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit