Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KLF7 Antibody (3G3), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP242626FR
Description
KLF7 Monoclonal specifically detects KLF7 in Human samples. It is validated for Western Blot, ELISA.Specifications
KLF7 | |
Monoclonal | |
DyLight 680 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
KLF7 | |
Recombinant human KLF7 (1-302aa) purified from E. coli | |
100 μL | |
Primary | |
Human | |
Purified |
Western Blot, ELISA | |
3G3 | |
Western Blot, ELISA | |
Krueppel-like factor 7, Kruppel-like factor 7 (ubiquitous), ubiquitous Kruppel-like transcription factor, UKLFUbiquitous krueppel-like factor | |
Mouse | |
Protein A purified | |
RUO | |
8609 | |
Store at 4C in the dark. | |
IgG1 κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction