Learn More
Aviva Systems Biology Corp KLF8 Antibody - N-terminal region (ARP57952_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP57952_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KLF8. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human KLF8. Alternate names: BKLF3, DKFZp686O08126, DXS741, MGC138314, ZNF741ApplicationWB. Species reactivity: Guinea Pig, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-62188