Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KLF8 Polyclonal Antibody
KLF8 Polyclonal Antibody
Supplier: Thermo Scientific PA5109885
Description
KLF8 Polyclonal Antibody for Western Blot, ICC/IF

Specifications
KLF8 | |
Polyclonal | |
Unconjugated | |
KLF8 | |
A830097P10Rik; Basic krueppel-like factor 3; basic kruppel-like factor 3; BKLF3; DKFZp686O08126; DXS741; KLF8; Klf8-ps1; Krueppel-like factor 8; Kruppel like factor 8; Kruppel-like factor 8; Kruppel-like factor 8, pseudogene 1; LOW QUALITY PROTEIN: Krueppel-like factor 8; MGC138314; RGD1565831; RP13-1021K9.1; zinc finger protein 741; ZNF74; ZNF741 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
11279, 245671, 302582 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
0.35 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
O95600, Q8BLM0 | |
KLF8 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 210-280 of human KLF8 (NP_009181.2). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction