Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KLF9 Mouse anti-Human, Janelia Fluor™ 549, Clone: OTI8A11, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270285JF549

Catalog No. N270285J549


Add to cart

Description

Description

KLF9 Monoclonal antibody specifically detects KLF9 in Human samples. It is validated for Immunohistochemistry.
Specifications

Specifications

KLF9
OTI8A11
Immunohistochemistry
Affinity Purified, Conjugated
KLF9
Full length recombinant protein of human KLF9 (NP_001197) produced in E.coli.
Immunogen affinity purified
Store at 4°C in the dark.
Monoclonal
Human
Immunohistochemistry
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
basic transcription element binding protein 1, Basic transcription element-binding protein 1, BTEB1GC-box-binding protein 1, BTEBBTE-binding protein 1, Krueppel-like factor 9, Kruppel-like factor 9, Transcription factor BTEB1
Mouse
IgG1
0.1 mL
Primary
687.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit