Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KLF9 Rabbit anti-Human, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$106.19 - $320.71

Specifications

Antigen KLF9
Dilution Western Blot 1.0 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Conjugate Unconjugated
Format Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB126699
View Documents
Novus Biologicals
NBP310899100UL
100 μg
Each for $320.71
Add to cart
 
NB126700
View Documents
Novus Biologicals
NBP31089925UL
25 μg
Each for $106.19
Add to cart
 
Description

Description

KLF9 Polyclonal antibody specifically detects KLF9 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications

Specifications

KLF9
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Purified
RUO
PBS buffer, 2% sucrose
687
Polyclonal
Affinity purified
Western Blot 1.0 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin
Unconjugated
Rabbit
Human, Rat
basic transcription element binding protein 1, Basic transcription element-binding protein 1, BTEB1GC-box-binding protein 1, BTEBBTE-binding protein 1, Krueppel-like factor 9, Kruppel-like factor 9, Transcription factor BTEB1
The immunogen is a synthetic peptide directed towards the N-terminal region of Human KLF9 (NP_001197). Peptide sequence LPEREVTKEHGDPGDTWKDYCTLVTIAKSLLDLNKYRPIQTPSVCSDSLE
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit