Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ KLH 45

Potent DDHD2 inhibitor

$163.90 - $686.50

Specifications

CAS 1632236-44-2
Formula Weight 458.48g/mol
For Use With (Application) Hydrolase Inhibitors
Molecular Formula C24H25F3N4O2
Storage Requirements Store at −20°C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
58-651-0 View Documents Tocris Bioscience™
5865/10
10mg Each for $163.90
Add to cart
 
58-655-0 View Documents Tocris Bioscience™
5865/50
50mg Each for $686.50
Add to cart
 
Description

Description

Potent DDHD2 inhibitor (IC50 = 1.3 nM). Selective for DDHD2 over 40 other serine hydrolases, but does exhibit cross-reactivity with ABHD6. Active in vivo. Increases brain triacylglycerol levels in mice.
Specifications

Specifications

1632236-44-2
Hydrolase Inhibitors
Store at −20°C
458.48g/mol
C24H25F3N4O2
Hydrolase

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit