Learn More
Aviva Systems Biology Corp KLHDC2 ANTIBODY 50UG/50UL
Supplier:  Aviva Systems Biology Corp ARP55015_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KLHDC2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human KLHDC2. Alternate names: HCLP-1, LCPApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-61397
Item Discontinued This item has been discontinued and is no longer available. Please reach out to your sales representative.