Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KLHDC2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA590252
Description
Positive Samples: rat pancreas; Cellular Location: Nucleus Immunogen sequence: EWIQFNHPYT EKPRLWHTAC ASDEGEVIVF GGCANNLLVH HRAAHSNEIL IFSVQPKSLV RLSLEAVICF KEMLANSWNC LPKHLLHSVN QRFGSNNTSG S
Represses CREB3-mediated transcription by interfering with CREB3-DNA binding. [UniProt]
Specifications
KLHDC2 | |
Polyclonal | |
Unconjugated | |
Klhdc2 | |
2310022K15Rik, D12Ertd522e, endothelial differentiation inhibitory protein TNG, HCA33, HCLP1, HCLP-1, Hepatocellular carcinoma-associated antigen 33, host cell factor homolog LCP, Host cell factor-like protein 1, kelch domain containing 2, kelch domain-containing protein 2, KLHDC2, LCP, similar to kelch domain containing 2 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
23588, 299113, 69554 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
1.46 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300, pH 7.3 | |
Q3KRE6, Q4G5Y1, Q9Y2U9 | |
Klhdc2 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 306-406 of human KLHDC2 (NP_0551301) | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction