Learn More
KLHDC4 Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 169151AP
Catalog No. 50-173-1256
Add to cart
Specifications
Specifications
KLHDC4
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8TBB5, Q921I2
kelch domain containing 4, KLHDC4
Rabbit
IgG
150 μL
-20°C
Primary
234825, 54758
Western Blot
Unconjugated
KLHDC4
Liquid
KLHDC4
KLHDC4 Fusion Protein Ag10096
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse