Learn More
Invitrogen™ KLHL12 Monoclonal Antibody (2G2)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA515649
Description
MA5-15649 targets KLHL12 in indirect ELISA, WB applications and shows reactivity with Human samples. The MA5-15649 immunogen is purified recombinant fragment of human KLHL12 expressed in E. Coli. MA5-15649 detects KLHL12 which has a predicted molecular weight of approximately 63kDa.
Serves as a substrate-specific adapter for the CUL3-based ubiquitin-protein E3 ligase complex. Negatively regulates the Wnt signaling pathway via the targeted ubiquitination and subsequent proteolysis of DVL3.Tissue specificity: Highly expressed in testis and at lower levels in the submandibular salivary gland.
Specifications
KLHL12 | |
Monoclonal | |
Conc. Not Determined | |
ascites with 0.03% sodium azide | |
Q53G59 | |
Klhl12 | |
Purified recombinant fragment of human KLHL12 expressed in E. Coli. | |
RUO | |
59349 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Western Blot | |
2G2 | |
Unconjugated | |
Klhl12 | |
C3IP1; CUL3-interacting protein 1; DKIR; DKIR homolog; FLJ27152; hDKIR; kelch like family member 12; kelch-like 12; kelch-like family member 12; kelch-like protein 12; kelch-like protein C3IP1; Klhl12; si:dkeyp-53e12.5; Unknown (protein for MGC:134560); wu:fi60g03; zgc:92570 | |
Mouse | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.