Learn More
Aviva Systems Biology Corp KLHL15 Antibody - middle region (ARP58487_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP58487_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KLHL15. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human KLHL15. Alternate names: MGC126148, MGC126149ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat, Zebrafish
Catalog No. 50-014-62321