Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KLHL15 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA586000
Description
KLHL15 Polyclonal Antibody for Western Blot, ICC/IF
KLHL15 is a member of a growing superfamily of kelch repeat-containing proteins. The kelch motif was discovered in the sequence of the Drosophila kelch ORF1 protein and forms a conserved beta-propeller tertiary structure. These kelch-like proteins have been implicated in embryogenesis and carcinogenesis through cytoskeleton organization. KLHL15 mRNA is widely expressed in many tissues and is thought to be involved in protein ubiquination and cytoskeleton organization.
Specifications
KLHL15 | |
Polyclonal | |
Unconjugated | |
KLHL15 | |
6330500C13Rik; epididymis secretory protein Li 305; HEL-S-305; kelch like family member 15; kelch-like 15; kelch-like family member 15; kelch-like protein 15; KIAA1677; KLHL15; RGD1563101 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
236904, 314111, 80311 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 0.2% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4 | |
A2AAX3, D3ZA50, Q96M94 | |
KLHL15 | |
Synthetic peptide within Human KLHL15 aa 555-604. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction