Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KLHL2 Mouse anti-Human, Canine, Monkey, PE-Cyanine7, Clone: OTI1G7, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271695PECY7

Catalog No. N271695PEC7


Add to cart

Description

Description

KLHL2 Monoclonal antibody specifically detects KLHL2 in Human, Canine, Monkey samples. It is validated for Flow Cytometry.
Specifications

Specifications

KLHL2
OTI1G7
Flow Cytometry
Affinity Purified, Conjugated
KLHL2
Recombinant protein fragment corresponding to amino acids 1-100 and 494-593 of human KLHL2 (NP_009177) produced in HEK293T cells.
Immunogen affinity purified
Store at 4°C in the dark. Do not freeze.
Monoclonal
Canine, Human, Monkey
Flow Cytometry
PE-Cyanine7
PBS with 0.05% Sodium Azide
ABP-KELCH, Actin-binding protein Mayven, kelch, kelch (Drosophila)-like 2 (Mayven), kelch-like 2, Mayven (Drosophila), kelch-like protein 2, MAV, MAYVEN
Mouse
IgG2b
0.1 mL
Primary
11275.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit