Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KLHL2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA550436

Catalog No. PA550436


Add to cart

Description

Description

The antibody detects endogenous levels of total KLHL2 protein.

KLHL2 (kelch-like protein 2), also known as actin-binding protein Mayven, is a 593 amino acid actin-binding protein. Highly expressed in brain, KLHL2 is thought to play a role in the organization of the actin cytoskeleton in brain cells.
Specifications

Specifications

KLHL2
Polyclonal
Unconjugated
KLHL2
Liquid
KLHL2
Fusion protein corresponding to a region derived from internal residues of human kelch-like family member 2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin), Western Blot
0.9 mg/mL
PBS with 50% glycerol and 0.05% sodium azide; pH 7.3
O95198
MAV, MAYVEN, ABP-KELCH
Rabbit
IgG
100 μL
-20°C
Primary
11275
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit