Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KLHL23 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA569117
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
KLHL23 is a protein coding gene.
Specifications
KLHL23 | |
Polyclonal | |
Unconjugated | |
KLHL23 | |
C130068N17Rik; DITHP; kelch like family member 23; kelch-like 23; kelch-like family member 23; kelch-like protein 23; Klhl23; RGD1307166 | |
Rabbit | |
Affinity chromatography | |
RUO | |
151230 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8NBE8 | |
KLHL23 | |
synthetic peptide directed towards the middle region of human KLHL23. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction