Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KLHL35 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP17955020UL

 View more versions of this product

Catalog No. NBP17955020


Add to cart

Description

Description

KLHL35 Polyclonal antibody specifically detects Antigen in Human samples. It is validated for Western Blotting.
Specifications

Specifications

KLHL35
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
Affinity Purified
KLHL35
Synthetic peptide directed towards the N terminal of human FLJ33790The immunogen for this antibody is FLJ33790. Peptide sequence RPRRFMDLAEVIVVIGGCDRKGLLKLPFADAYHPESQRWTPLPSLPGYTR.
Immunogen affinity purified
RUO
Primary
283212
Western Blot
Western Blot 1:1000
NP_001034637
FLJ33790, kelch-like 35 (Drosophila)
Rabbit
IgG
20ul
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit