Learn More
Invitrogen™ KLK4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5109888
Description
Immunogen sequence: QIINGEDCSP HSQPWQAALV MENELFCSGV LVHPQWVLSA AHCFQNSYTI GLGLHSLEAD QEPGSQMVEA SLSVRHPEYN RPLLANDLML IKLDESVSES DTIRSISIAS QCPTAGNSCL VSGWGLLANG RMPTVLQCVN VSVVSEEVCS KLYDPLYHPS MFCAGGGQDQ KDSCNGDSGG PLICNGYLQG LVSFGKAPCG QVGVPGVYTN LCKFTEWIEK TVQAS.
Kallikreins are a subgroup of serine proteases and are implicated in carcinogenesis. A novel Kallikrein gene Prostase/KLK-L1 (also known as KLK4) is expressed and is up-regulated by androgens and progestins. KLK-L1 may be involved in the pathogenesis and/or progression of prostate, breast, and possibly other malignancies.
Specifications
KLK4 | |
Polyclonal | |
Unconjugated | |
KLK4 | |
AI2A1; androgen-regulated message 1; ARM1; EMSP; Emsp1; enamel matrix serine protease 1; Enamel matrix serine proteinase 1; ESMP1; kallikrein; kallikrein 4 (prostase, enamel matrix, prostate); kallikrein related peptidase 4; kallikrein related-peptidase 4 (prostase, enamel matrix, prostate); kallikrein-4; Kallikrein-like protein 1; kallikrein-related peptidase 4; Klk4; KLK-4; KLKL1; KLK-L1; Prostase; protease, serine 17 (enamel matrix, prostate); Prss17; PSTS; serine protease 17 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
56640, 9622 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
0.52 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
Q9Y5K2, Q9Z0M1 | |
KLK4 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 30-254 of human KLK4 (NP_004908.4). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.