Learn More
KLK8 Mouse anti-Human, Clone: OTI1G9, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF801720
Catalog No. 50-166-5701
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Kallikreins are a subgroup of serine proteases having diverse physiological functions. Growing evidence suggests that many kallikreins are implicated in carcinogenesis and some have potential as novel cancer and other disease biomarkers. This gene is one of the fifteen kallikrein subfamily members located in tandem in a gene cluster on chromosome 19. The encoded protein may be involved in proteolytic cascade in the skin and may serve as a biomarker for ovarian cancer. Alternate splicing of this gene results in multiple transcript variants encoding different isoforms.
Specifications
Specifications
KLK8
Monoclonal
Unconjugated
KLK8
Lyophilized
KLK8
Human recombit protein fragment corresponding to amino acids 33-260 of human KLK8 produced in SF9 cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Western Blot
OTI1G9
PBS with 8% trehalose and no preservative; pH 7.3
O60259
HNP, NP, NRPN, PRSS19, TADG14
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
11202