Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Klotho Monoclonal Antibody (C3), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA543727
This item is not returnable.
View return policy
Description
Klotho Monoclonal antibody specifically detects Klotho in Human, Rat samples. It is validated for Western Blot.Specifications
Klotho | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
Q9UEF7, Q9Z2Y9 | |
KL | |
Phe765-Ser981 (Accession Q9UEF7), with Two N-terminal Tags, His-tag and SUMO-tag | |
20 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG1 κ |
Western Blot | |
C3 | |
Unconjugated | |
KL | |
alpha-kl; HFTC3; Kl; Klotho; Klotho peptide; secreted form of Klotho protein | |
Mouse | |
Protein A/G | |
RUO | |
83504, 9365 | |
-20°C | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction