Learn More
Klotho Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA588303
Description
Immunogen sequence: MPASAPPRRP RPPPPSLSLL LVLLGLGGRR LRAEPGDGAQ TWARFSRPPA PEAAGLFQGT FPDGFLWAVG SAAYQTEGGW QQHGKGASIW DTFTHHPLAP PGDSRNASLP LGAPSPLQPA TGDVASDSYN NVFRDTEALR ELGVTHYRFS ISWARVLPNG SAGVPNREGL RYYRRLLERL RELGVQPVVT LYHWDLPQRL; Positive Samples: DU145, Mouse kidney, Mouse brain, Rat kidney; Cellular Location: Cell membrane, Single-pass type I membrane protein
Klotho is a novel protein, found in several species. It is important for it′s activity as a hormone, circulating through the blood and binding to cells. Therapies based on klotho could eventually prove to be a way to extend life or slow its effects. The Klotho gene seems to delay many of the effects of old age, such as the weakening of bones, clogging of the arteries and loss of muscle fitness.Specifications
Klotho | |
Polyclonal | |
Unconjugated | |
KL | |
Kl | |
Recombit fusion protein containing a sequence corresponding to amino acids 1-200 of human KL | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Immunofluorescence, Immunohistochemistry, Western Blot | |
2.86 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O35082, Q9UEF7, Q9Z2Y9 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
16591, 83504, 9365 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.