Learn More
Abnova™ KLRC2 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00003822T01
Catalog No. 89-950-131
Add to cart
Specifications
Specifications
293T
BC093644.1
pCMV-KLRC2 full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Western Blot
KLRC2 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 26.1kDa
Transient overexpression cell lysate was tested with anti-FASTK antibody (Mfr. No. H00003822-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Human