Learn More
KLRG1 Hamster anti-Mouse, BV421, Clone:  2F1, BD Horizon™
Hamster Monoclonal Antibody
Manufacturer:  BD Biosciences 566284
Catalog No. BDB566284
Add to cart
Description
Description
KLRG1 Monoclonal antibody specifically detects KLRG1 in Mouse samples. It is validated for Flow cytometry.
Specifications
Specifications
KLRG1
 2F1
Brilliant Violet 421
Purified
Hamster
Syrian Hamster IgG2, κ
RUO
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BV421 under optimum conditions, and unconjugated antibody and free BD Horizon™ BV421 were removed.
Monoclonal
Flow Cytometry
0.2mg/mL
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Klrg1; Killer cell lectin-like receptor subfamily G member 1; MAFA
A-LAK from C57BL/6 mice
25μg
Cell Biology
Primary
Murine
SDS
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Reseach Use Only.