Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KLRG1 Mouse anti-Human, Super Bright 702, Clone: 13F12F2, eBioscience™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

$161.00 - $342.00

Specifications

Antigen KLRG1
Clone 13F12F2
Target Species Human
Conjugate Super Bright 702
Isotype IgG2a, kappa
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
67948841 Non-distribution product as customer accommodation. View Documents Invitrogen
67948841
25 Tests Each for $161.00
Add to cart
 
67948842 Non-distribution product as customer accommodation. View Documents Invitrogen
67948842
100 Tests Each for $342.00
Add to cart
 
Description

Description

Description: The GIGD7 monoclonal antibody reacts with mouse T cell immunoreceptor with Ig and ITIM domains (TIGIT, VSTM3, WUCAM), a 26 kDa protein that is a member of the poliovirus receptor (PVR) family. In mice, the expression of TIGIT has been reported in follicular T helper cells, while in humans, the expression of TIGIT has been reported on NK cells, regulatory T cells, follicular T helper cells, memory CD4+ T cells, and CD8+ T cells. TIGIT is not expressed on B cells or naive CD4+ T cells. TIGIT is upregulated on CD4+ T cells following activation. TIGIT can interact with certain members of the PVR and PVR-like families, including CD155, and also mediates the interaction of NK cells and T cells with antigen presenting cells, fibroblasts and endothelial cells that express PVR and PVR-like proteins. Applications Reported: This GIGD7 antibody has been reported for use in flow cytometric analysis. Applications Tested: This GIGD7 antibody has been tested by flow cytometric analysis of mouse splenocytes. This may be used at less than or equal to 1.0 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

Super Bright 702 is a tandem dye that can be excited with the violet laser line (405 nm) and emits at 702 nm. We recommend using a 710/50 bandpass filter. Please make sure that your instrument is capable of detecting this fluorochrome. When using two or more Super Bright dye-conjugated antibodies in a staining panel, it is recommended to use Super Bright Staining Buffer (Product No. SB-4400) to minimize any non-specific polymer interactions. Please refer to the datasheet for Super Bright Staining Buffer for more information. Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (Product No. 00-8222) (100 µL of cell sample + 100 µL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (Product No. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 405 nm; Emission: 702 nm; Laser: Violet Laser KLRG1 (Killer cell lectin-like receptor G1, MAFA, 2F1-Ag) is a homodimeric member of the lectin-like type 2 transmembrane receptor family whose members contain characteristic immunoreceptor tyrosine-based inhibitory motifs (ITIMs) in their cytoplasmic domains. These ITIMs interact with the SH2 domains of protein phosphatases such as SHP-1. Functionally, KLRG1 is a lymphocyte that is involved in the lysis of certain tumor cells and virus-infected cells, and mediate humoral and cell-mediated immune responses.
Specifications

Specifications

KLRG1
Human
IgG2a, kappa
Monoclonal
Liquid
Antibody
4° C, store in dark, DO NOT FREEZE!
13F12F2
Super Bright 702
Flow Cytometry
Primary
Monoclonal
PBS with BSA and 0.09% sodium azide; pH 7.2
RUO

For Research Use Only

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit