Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KLRG2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548757

Catalog No. PA548757

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

KLRG2
Polyclonal
PBS with 0.09% sodium azide
A4D1S0
Killer cell lectin-like receptor subfamily G member 2, C-type lectin domain family 15 member B, KLRG2, CLEC15B
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
346689
Western Blot
Unconjugated
KLRG2
Liquid
KLRG2
KLH conjugated synthetic peptide between 20-49 amino acids from the N-terminal region of human KLRG2.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit