KM 11060, Tocris

Corrects F508del-CFTR trafficking

Manufacturer: R&D Systems 3143/50

Quantity 50mg

 View more versions of this product

Catalog No. 31-435-0

  Check Availability
Add to Cart

    Chemical Identifiers

    Assay Percent Range >98%
    Molecular Formula C19H17Cl2N3O2S
    Formula Weight 422.33
    Synonym 7-Chloro-4-[4-[4-chlorophenyl)sulfonyl]-1-piperazinyl]quinoline
    Chemical Name or Material KM 11060

    Specifications

    Formula Weight 422.33
    Quantity 50mg
    Assay Percent Range >98%
    Molecular Formula C19H17Cl2N3O2S
    CAS 774549-97-2
    Synonym 7-Chloro-4-[4-[4-chlorophenyl)sulfonyl]-1-piperazinyl]quinoline
    Recommended Storage Store at Room Temperature
    Chemical Name or Material KM 11060

    Store at Room Temperature

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit