Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KM ACT Cleanroom Paper Non-distribution product as customer accommodation.

Sky blue paper for writing, printing or photocopying in a cleanroom environment

$109.29 - $117.79

Specifications

Color Sky Blue
For Use With (Application) For writing, printing or photocopying in a cleanroom environment
Product Type Cleanroom Paper
View More Specs

Products 2
Catalog Number Mfr. No. Weight (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Weight (Metric) Price Quantity & Availability  
17-607-308 Non-distribution product as customer accommodation.
View Documents
KM ACT
KMCPS72BLT
75g
Case of 2500 for $109.29
Add to cart
 
17-607-620 Non-distribution product as customer accommodation.
View Documents
KM ACT
KMCP80BLT
80g
Case of 2500 for $117.79
Add to cart
 
Description

Description

  • Class 1 and other cleanroom use
  • Available in two weights

Specifications

Specifications

Sky Blue
For writing, printing or photocopying in a cleanroom environment
Cleanroom Paper
SDS
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit