Learn More
KNF™ FLEXPAK Antistatic Cleanroom Bags
Constructed from low-density amine-free polyethylene film
Supplier: KNF™ FLEXPAK 020500914
Description
Flat construction.
- 6mil thick
- Tough yet flexible
- Exhibits low water vapor transmission rate and good electrical properties
Certifications
Meets NASA JPG 5322.1 Level 150. Certificate of conformance accompanies shipment and reflects part number and lot number for traceability.
Compliance
Suitable for ISO Class 5 cleanroom environments
Specifications
NASA JPG 5322.1 Level 150. Certificate of conformance accompanies shipment and reflects part number | |
6 mil | |
120 in. | |
120 in. |
Polyethylene | |
120 x 120 in. (305 x 305 cm) | |
305 cm | |
305 cm |