Learn More
KNF™ FLEXPAK Antistatic Cleanroom Bags
Constructed from low-density amine-free polyethylene film
$26.42 - $107.64
Specifications
Certifications/Compliance NASA JPG 5322.1 Level 150. Certificate of conformance accompanies shipment and reflects part number
Material PE (Polyethylene)
Thickness (English) 6 mil
View More Specs
Products 14
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity & Availability  
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity & Availability  
19-130-2147
View Documents
KNF™ FLEXPAK
020500706
60 x 48 in. (153 x 122cm)
Each for $29.64
Add to cart
19-130-2150
View Documents
KNF™ FLEXPAK
020500710
72 x 60 in. (183 x 153cm)
Each for $36.71
Add to cart
19-130-2155
View Documents
KNF™ FLEXPAK
020500842
96 x 84 in. (244 x 214cm)
Each for $66.35
Add to cart
19-130-2156
View Documents
KNF™ FLEXPAK
020500843
96 x 96 in. (244 x 244cm)
Each for $74.26
Add to cart
19-130-2162
View Documents
KNF™ FLEXPAK
020500966
156 x 96 in. (397 x 244cm)
Each for $102.23
Add to cart
19-130-2161
View Documents
KNF™ FLEXPAK
020500951
132 x 108 in. (335 x 274cm)
Each for $107.64
Add to cart
19-130-2146
View Documents
KNF™ FLEXPAK
020500700
48 x 48 in. (122 x 122cm)
Each for $29.38
Add to cart
19-130-2148
View Documents
KNF™ FLEXPAK
020500707
60 x 60 in. (153 x 153cm)
Each for $34.53
Add to cart
19-130-2149
View Documents
KNF™ FLEXPAK
020500709
72 x 48 in. (183 x 122cm)
Each for $34.53
Add to cart
19-130-2154
View Documents
KNF™ FLEXPAK
020500828
96 x 72 in. (244 x 183cm)
Each for $57.72
Add to cart
19-130-2198
View Documents
KNF™ FLEXPAK
020500817
96 x 60 in. (244 x 153cm)
Each for $33.75
Add to cart
19-130-2159
View Documents
KNF™ FLEXPAK
020500907
36 x 36 in. (92 x 92cm)
Each for $26.42
Add to cart
19-130-2158
View Documents
KNF™ FLEXPAK
020500906
120 x 72 in. (305 x 183cm)
Each for $72.90
Add to cart
19-130-2200
View Documents
KNF™ FLEXPAK
020500914
120 x 120 in. (305 x 305cm)
Each for $33.51
Case of 20 EA for $1,954.16
Add to cart
Description
Description
Flat construction.
  • 6mil thick
  • Tough yet flexible
  • Exhibits low water vapor transmission rate and good electrical properties
Certifications
Meets NASA JPG 5322.1 Level 150. Certificate of conformance accompanies shipment and reflects part number and lot number for traceability.
Compliance
Suitable for ISO Class 5 cleanroom environments
Specifications
Specifications
NASA JPG 5322.1 Level 150. Certificate of conformance accompanies shipment and reflects part number
PE (Polyethylene)
6 mil