KNF™ FLEXPAK Antistatic Cleanroom Sheeting

Easily heat-sealed for bag manufacture

Manufacturer: KNF Flexpak Corporation 020501841

 View more versions of this product

Catalog No. 19-168-089

  Check Availability
Add to Cart

    Specifications

    Certifications/Compliance NASA JPG 5322.1 Level 150. Certificate of conformance accompanies shipment and reflects part number and lot number for traceability; Suitable for ISO Class 5 cleanroom environments
    Material Polyethylene
    Length (English) 96 in.
    Width (English) 96 in.
    Width (Metric) 244cm
    Thickness (English) 6 mil
    Dimensions (L x W) 96 x 96 in. (244 x 244cm)
    Length (Metric) 244cm

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit