Learn More
KNF FLEXPAK Antistatic Cleanroom Sheeting
Easily heat-sealed for bag manufacture
$10.79 - $190.30
Specifications
Certifications/Compliance | NASA JPG 5322.1 Level 150. Certificate of conformance accompanies shipment and reflects part number |
---|---|
Material | PE (Polyethylene) |
Thickness (English) | 6 mil |
Catalog Number | Mfr. No. | Dimensions (L x W) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Dimensions (L x W) | Price | Quantity & Availability | |||||
19-130-2145
|
KNF FLEXPAK
020500664 |
36 x 24 in. (92 x 61cm) |
Each for $25.40
|
|
19-168-071
|
KNF FLEXPAK
020500708 |
72 x 36 in. (183 x 92cm) |
Each for $29.55
|
|
19-130-2152
|
KNF FLEXPAK
020500738 |
84 x 48 in. (213 x 122cm) |
Each for $34.25
|
|
19-168-072
|
KNF FLEXPAK
020500789 |
96 x 48 in. (244 x 122cm) |
Each for $10.79
|
|
19-168-075
|
KNF FLEXPAK
020500909 |
156 x 120 in. (397 x 305cm) |
Each for $87.90
|
|
19-168-073
|
KNF FLEXPAK
020500855 |
108 x 72 in. (275 x 183cm) |
Each for $44.70
|
|
19-168-082
|
KNF FLEXPAK
020501291 |
48 x 48 in. (122 x 122cm) |
Each for $17.05
|
|
19-168-084
|
KNF FLEXPAK
020501293 |
72 x 60 in. (183 x 153cm) |
Each for $87.90
|
|
19-168-088
|
KNF FLEXPAK
020501473 |
60 x 60 in. (153 x 153cm) |
Each for $42.10
|
|
19-168-086
|
KNF FLEXPAK
020501295 |
48 x 36 in. (122 x 92cm) |
Each for $20.99
|
|
19-168-085
|
KNF FLEXPAK
020501294 |
60 x 60 in. (153 x 153cm) |
Each for $42.10
|
|
19-168-092
|
KNF FLEXPAK
020501844 |
108 x 108 in. (275 x 275cm) |
Each for $83.70
|
|
19-168-068
|
KNF FLEXPAK
020500519 |
144 x 120 in. (366 x 305cm) |
Each for $109.80
Case of 10 EA for $979.50 |
|
19-130-2197
|
KNF FLEXPAK
020500695 |
48 x 36 in. (122 x 92cm) |
Each for $19.60
|
|
19-130-2199
|
KNF FLEXPAK
020500846 |
108 x 60 in. (275 x 153cm) |
Each for $53.40
|
|
19-168-089
|
KNF FLEXPAK
020501841 |
96 x 96 in. (244 x 244cm) |
|
19-130-2153
|
KNF FLEXPAK
02050788 |
84 x 72 in. (213 x 183cm) |
Each for $54.40
|
|
19-168-091
|
KNF FLEXPAK
020501843 |
168 x 144 in. (427 x 366cm) |
Each for $190.30
Case of 10 EA for $1,715.00 |
|
19-168-090
|
KNF FLEXPAK
020501842 |
156 x 120 in. (397 x 305cm) |
Each for $152.70
|
|
19-168-093
|
KNF FLEXPAK
020501845 |
92 x 92 in. (244 x 244cm) |
Each for $91.00
|
|
Description
Constructed from low-density amine-free polyethylene film. Flat construction.
- 6mil thick sheeting
- Tough yet flexible
- Exhibits low water vapor transmission rate and good electrical properties
Certifications
Meets NASA JPG 5322.1 Level 150. Certificate of conformance accompanies shipment and reflects part number and lot number for traceability.
Compliance
Suitable for ISO Class 5 cleanroom environments
Specifications
NASA JPG 5322.1 Level 150. Certificate of conformance accompanies shipment and reflects part number | |
PE (Polyethylene) | |
6 mil |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.