Learn More
KNF™ FLEXPAK Antistatic Polyethylene Clear Tubing
Manufacturer:  KNF™ FLEXPAK 020500289
 View more versions of this product
Catalog No. 19-168-095
Add to cart
Description
Description
  • Polyethylene (PE) / Cross-linked Polyethylene (PEX) material
  • Chemically resistant to solvents
  • Good electrical properties
  • Puncture-resistant
  • Low density
  • Thickness: 6 mil
Specifications
Specifications
Effective for secondary packaging for parts transport
500 ft.
10 in.
Polyethylene