Learn More
KNF™ FLEXPAK Polyethylene Ultraclean™ Bags
Low-density polyethylene films are proven performers
Manufacturer:  KNF™ FLEXPAK LB1041010
 View more versions of this product
Catalog No. 18-063-578
Add to cart
Description
Description
  • Tough yet flexible
  • Easily heat-sealed for bag manufacture
  • Strong puncture-resistance
  • Low water-vapor transmission rate
  • Good electrical properties
  • Amine-free anti-static agent
  • Chemically resistant to most solvents
Certifications
FDA approved for use with food
Specifications
Specifications
10 x 8 in. (25.4 x 20.3cm)
4 mil
25.4cm
25.4cm
10 in.
8 in.
PE (Polyethylene)