Learn More
KNF™ FLEXPAK Square-Bottom Antistatic Cleanroom Bags
SDP
Square-bottom, antistatic bags are treated with an amine-free agent
$7.65 - $1264.00
Specifications
Material Polyethylene
View More Specs
Products 33
Catalog Number Mfr. No. Height (English) Dimensions (L x W) Price Quantity & Availability  
Catalog Number Mfr. No. Height (English) Dimensions (L x W) Price Quantity & Availability  
19-130-2132
SDP
View Documents
KNF™ FLEXPAK
020500359
40 in. 28 x 32 in. (71.12 x 81.3 cm)
Each for $97.00
Add to Cart
19-167-960
SDP
View Documents
KNF™ FLEXPAK
020500365
44 in. 24 x 44 in. (60.96 x 111.8 cm)
Each for $101.00
Add to Cart
19-168-008
SDP
View Documents
KNF™ FLEXPAK
020500552
44 in. 72 x 88 in. (182.88 x 223.5 cm)
Each for $172.00
Add to Cart
19-167-951
SDP
View Documents
KNF™ FLEXPAK
020500012
48 in. 24 x 24 in. (60.96 x 61 cm)
Each for $83.00
Add to Cart
19-167-950
SDP
View Documents
KNF™ FLEXPAK
020500001
36 in. 24 x 24 in. (60.96 x 61 cm)
Each for $28.00
Add to Cart
19-167-953
SDP
View Documents
KNF™ FLEXPAK
020500217
40 in. 24 x 28 in. (60.96 x 71 cm)
Each for $70.75
Add to Cart
19-167-954
SDP
View Documents
KNF™ FLEXPAK
020500221
44 in. 24 x 32 in. (60.96 x 81.3 cm)
Each for $89.00
Add to Cart
19-130-2131
SDP
View Documents
KNF™ FLEXPAK
020500220
32 x 24 in. (81.28 x 61 cm)
Each for $99.00
Add to Cart
19-130-2133
SDP
View Documents
KNF™ FLEXPAK
020500363
36 in. 40 x 24 in. (101.6 x 61 cm)
Each for $97.00
Add to Cart
19-167-961
SDP
View Documents
KNF™ FLEXPAK
020500367
48 in. 24 x 48 in. (60.96 x 121.9 cm)
Each for $126.00
Add to Cart
19-167-962
SDP
View Documents
KNF™ FLEXPAK
020500370
44 in. 24 x 44 in. (60.96 x 111.8 cm)
Each for $101.00
Add to Cart
19-167-963
SDP
View Documents
KNF™ FLEXPAK
020500372
32 in. 20 x 40 in. (50.8 x 101.6 cm)
Each for $83.00
Add to Cart
19-167-964
SDP
View Documents
KNF™ FLEXPAK
020500374
44 in. 28 x 44 in. (71.12 x 111.8 cm)
Each for $106.00
Add to Cart
19-167-970
SDP
View Documents
KNF™ FLEXPAK
020500386
44 in. 40 x 40 in. (101.6 x 101.6 cm)
Each for $108.00
Add to Cart
19-167-984
SDP
View Documents
KNF™ FLEXPAK
020500414
36 in. 24 x 32 in. (60.96 x 81.3 cm)
Each for $91.00
Add to Cart
19-167-983
SDP
View Documents
KNF™ FLEXPAK
020500412
36 in. 20 x 36 in. (50.8 x 91.4 cm)
Each for $22.86
Add to Cart
19-167-998
SDP
View Documents
KNF™ FLEXPAK
020500475
36 in. 24 x 28 in. (60.96 x 71 cm)
Each for $83.00
Add to Cart
19-130-2140
SDP
View Documents
KNF™ FLEXPAK
020500594
48 in. 28 x 24 in. (71.12 x 61 cm)
Each for $106.00
Add to Cart
19-168-011
SDP
View Documents
KNF™ FLEXPAK
020500616
32 in. 13 x 14 in. (33.02 x 35.6 cm)
Each for $7.65
Add to Cart
19-168-012
SDP
View Documents
KNF™ FLEXPAK
020500617
48 in. 13 x 15 in. (33.02 x 38.1 cm)
Each for $8.79
Add to Cart
19-168-014
SDP
View Documents
KNF™ FLEXPAK
020500619
18 x 21 in. (45.72 x 53.3 cm)
Each for $54.75
Add to Cart
19-130-2170
SDP
View Documents
KNF™ FLEXPAK
020500792
32 in. 52 x 20 in. (132.08 x 50.8 cm)
Each for $95.00
Add to Cart
19-130-2175
SDP
View Documents
KNF™ FLEXPAK
020500797
96 in. 88 x 40 in. (223.5 x 101.6 cm)
Each for $175.00
Add to Cart
19-130-2184
SDP
View Documents
KNF™ FLEXPAK
020500854
40 in. 44 x 44 in. (111.76 x 111.8 cm)
Each for $71.25
Add to Cart
19-130-2190
SDP
View Documents
KNF™ FLEXPAK
020500919
40 in. 28 x 40 in. (71.12 x 101.6 cm)
Each for $82.00
Add to Cart
19-130-2143
SDP
View Documents
KNF™ FLEXPAK
020501129
48 in. 48 x 48 in. (121.92 x 121.9 cm)
Each for $143.00
Add to Cart
19-130-2212
SDP
View Documents
KNF™ FLEXPAK
020501646
52 in. 40 x 40 in. (101.6 x 101.6 cm)
Each for $157.00
Add to Cart
19-168-056
SDP
View Documents
KNF™ FLEXPAK
020501815
40 in. 20 x 24 in. (50.8 x 61 cm)
Each for $69.25
Add to Cart
19-168-058
SDP
View Documents
KNF™ FLEXPAK
020501817
48 in. 28 x 64 in. (71.12 x 162.6 cm)
Each for $111.00
Add to Cart
19-168-063
SDP
View Documents
KNF™ FLEXPAK
020535174
32 in. 28 x 29 in. (71.12 x 73.7 cm)
Each for $67.00
Add to Cart
19-165-344
SDP
View Documents
KNF™ FLEXPAK
020500359B
40 in. 28 x 32 in. (71.12 x 81.3 cm)
Pack for $806.00
Add to Cart
19-165-345
SDP
View Documents
KNF™ FLEXPAK
020500365B
44 in. 24 x 44 in. (60.96 x 111.8 cm)
Case of 30 for $1,264.00
Add to Cart
19-165-351
SDP
View Documents
KNF™ FLEXPAK
020500552B
44 in. 72 x 88 in. (182.88 x 223.5 cm)
Pack of 5 for $586.00
Add to Cart
Description
Description
Constructed from proven low-density polyethylene film. Perform well in a broad spectrum of applications. Ideal for parts that require a moderate level (Levels II and III) of packaging cleanliness.
  • Tough yet flexible
  • Chemically resistant to solvents
  • Exhibit low water vapor transmission rate and good electrical properties
Certifications
Cleaned and packaged in a Class 10/100 cleanroom, and certified to KNF Cleanroom Products Cleanliness Level II or III. FDA approved for use with food.
Compliance
Suitable for Class 100 cleanroom environments
Specifications
Specifications
Polyethylene