Learn More
KNF™ FLEXPAK KNF™ FLEXPAK Square-Bottom Antistatic Cleanroom Bags
Square-bottom, antistatic bags are treated with an amine-free agent
Manufacturer:  KNF™ FLEXPAK 020500364
 View more versions of this product
Catalog No. 19-167-959
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Constructed from proven low-density polyethylene film. Perform well in a broad spectrum of applications. Ideal for parts that require a moderate level (Levels II and III) of packaging cleanliness.
  • Tough yet flexible
  • Chemically resistant to solvents
  • Exhibit low water vapor transmission rate and good electrical properties
Certifications
Cleaned and packaged in a Class 10/100 cleanroom, and certified to KNF Cleanroom Products Cleanliness Level II or III. FDA approved for use with food.
Compliance
Suitable for Class 100 cleanroom environments
Specifications
Specifications
40 x 28 in. (101.6 x 71.12cm)
40 in.
101.6cm
71.12cm
80 in.
28 in.
PE (Polyethylene)