Learn More
KNF™ FLEXPAK Square-Bottom Antistatic Cleanroom Bags
Square-bottom, antistatic bags are treated with an amine-free agent
$59.80 - $575.64
Specifications
Material PE (Polyethylene)
View More Specs
Products 36
Catalog Number Mfr. No. Height (English) Length (English) Width (English) Price Quantity & Availability  
Catalog Number Mfr. No. Height (English) Length (English) Width (English) Price Quantity & Availability  
19-130-2191
View Documents
KNF™ FLEXPAK
020500921
60 in. 88 in. 84 in.
Each for $195.73
Case of 10 EA for $1,784.64
Add to cart
19-130-2144
View Documents
KNF™ FLEXPAK
020501168
64 in. 84 in. 120 in.
Each for $224.12
Add to cart
19-167-965
View Documents
KNF™ FLEXPAK
020500376
56 in. 44 in. 32 in.
Each for $69.06
Add to cart
19-167-968
View Documents
KNF™ FLEXPAK
020500382
60 in. 64 in. 520 in.
Each for $103.38
Add to cart
19-167-969
View Documents
KNF™ FLEXPAK
020500384
60 in. 36 in. 36 in.
Each for $84.86
Add to cart
19-167-974
View Documents
KNF™ FLEXPAK
020500394
60 in. 132 in. 88 in.
Each for $160.99
Add to cart
19-167-989
View Documents
KNF™ FLEXPAK
020500425
68 in. 152 in. 48 in.
Each for $196.87
Add to cart
19-167-994
View Documents
KNF™ FLEXPAK
020500467
52 in. 36 in. 28 in.
Each for $63.65
Add to cart
19-167-995
View Documents
KNF™ FLEXPAK
020500469
68 in. 72 in. 32 in.
Each for $87.05
Add to cart
19-167-997
View Documents
KNF™ FLEXPAK
020500473
64 in. 32 in. 24 in.
Each for $79.46
Add to cart
19-168-000
View Documents
KNF™ FLEXPAK
020500479
64 in. 108 in. 88 in.
Each for $142.48
Add to cart
19-130-2137
View Documents
KNF™ FLEXPAK
020500503
60 in. 48 in. 64 in.
Each for $120.74
Add to cart
19-165-348
View Documents
KNF™ FLEXPAK
020500503B
60 in. 64 in. 48 in.
Pack of 5 for $349.44
Add to cart
19-130-2138
View Documents
KNF™ FLEXPAK
020500505
64 in. 52 in. 72 in.
Each for $140.30
Add to cart
19-165-349
View Documents
KNF™ FLEXPAK
020500505B
64 in. 72 in. 52 in.
Pack of 5 for $413.40
Add to cart
19-168-006
View Documents
KNF™ FLEXPAK
020500507
68 in. 72 in. 72 in.
Each for $104.42
Add to cart
19-168-022
View Documents
KNF™ FLEXPAK
020500702
64 in. 44 in. 28 in.
Each for $71.86
Add to cart
19-168-023
View Documents
KNF™ FLEXPAK
020500704
52 in. 48 in. 44 in.
Each for $71.86
Add to cart
19-130-2180
View Documents
KNF™ FLEXPAK
020500848
52 in. 132 in. 52 in.
Each for $201.24
Add to cart
19-130-2181
View Documents
KNF™ FLEXPAK
020500849
52 in. 40 in. 36 in.
Each for $91.42
Add to cart
19-130-2186
View Documents
KNF™ FLEXPAK
020500865
56 in. 56 in. 20 in.
Each for $68.85
Case of 10 EA for $619.32
Add to cart
19-130-2189
View Documents
KNF™ FLEXPAK
020500918
52 in. 44 in. 24 in.
Each for $96.82
Add to cart
19-168-040
View Documents
KNF™ FLEXPAK
020500992
56 in. 36 in. 28 in.
Each for $59.80
Add to cart
19-168-041
View Documents
KNF™ FLEXPAK
020500993
68 in. 48 in. 44 in.
Each for $72.90
Add to cart
19-168-042
View Documents
KNF™ FLEXPAK
020500994
68 in. 52 in. 36 in.
Each for $88.09
Add to cart
19-168-053
View Documents
KNF™ FLEXPAK
020501480
66 in. 36 in. 20 in.
Each for $80.81
Add to cart
19-130-2213
View Documents
KNF™ FLEXPAK
020501645
68 in. 40 in. 28 in.
Each for $110.97
Add to cart
19-130-2211
View Documents
KNF™ FLEXPAK
020501659
60 in. 36 in. 32 in.
Each for $106.60
Add to cart
19-130-2210
View Documents
KNF™ FLEXPAK
020501660
64 in. 32 in. 32 in.
Each for $103.38
Add to cart
19-130-2224
View Documents
KNF™ FLEXPAK
020501662
60 in. 64 in. 60 in.
Each for $204.46
Add to cart
19-168-057
View Documents
KNF™ FLEXPAK
020501816
60 in. 32 in. 28 in.
Each for $60.94
Add to cart
19-168-059
View Documents
KNF™ FLEXPAK
020501823
64 in. 40 in. 40 in.
Each for $110.97
Add to cart
19-168-062
View Documents
KNF™ FLEXPAK
020535171
52 in. 28 in. 24 in.
Each for $62.61
Add to cart
19-165-350
View Documents
KNF™ FLEXPAK
020500545B
56 in. 32 in. 32 in.
Pack of 10 for $475.28
Add to cart
19-165-354
View Documents
KNF™ FLEXPAK
020500921B
60 in. 88 in. 84 in.
Pack of 5 for $575.64
Add to cart
19-165-357
View Documents
KNF™ FLEXPAK
020501168B
64 in. 120 in. 84 in.
Pack of 2 for $277.68
Add to cart
Description
Description
Constructed from proven low-density polyethylene film. Perform well in a broad spectrum of applications. Ideal for parts that require a moderate level (Levels II and III) of packaging cleanliness.
  • Tough yet flexible
  • Chemically resistant to solvents
  • Exhibit low water vapor transmission rate and good electrical properties
Certifications
Cleaned and packaged in a Class 10/100 cleanroom, and certified to KNF Cleanroom Products Cleanliness Level II or III. FDA approved for use with food.
Compliance
Suitable for Class 100 cleanroom environments
Specifications
Specifications
PE (Polyethylene)