Learn More
KNF™ FLEXPAK Square-Bottom Antistatic Cleanroom Bags
Square-bottom, antistatic bags are treated with an amine-free agent
$48.89 - $604.76
Specifications
Material PE (Polyethylene)
View More Specs
Products 44
Catalog Number Mfr. No. Height (English) Dimensions (L x W) Price Quantity & Availability  
Catalog Number Mfr. No. Height (English) Dimensions (L x W) Price Quantity & Availability  
19-168-004
View Documents
KNF™ FLEXPAK
020500504
80 in. 72 x 32 in. (182.88 x 81.28cm)
Each for $107.64
Add to cart
19-168-031
View Documents
KNF™ FLEXPAK
020500891
76 in. 128 x 84 in. (325.12 x 213.36cm)
Each for $219.96
Add to cart
19-167-955
View Documents
KNF™ FLEXPAK
020500303
76 in. 32 x 16 in. (81.28 x 40.64cm)
Each for $80.81
Add to cart
19-167-958
View Documents
KNF™ FLEXPAK
020500362
76 in. 40 x 20 in. (101.6 x 50.8cm)
Each for $76.96
Add to cart
19-167-966
View Documents
KNF™ FLEXPAK
020500378
80 in. 36 x 32 in. (91.44 x 81.28cm)
Each for $94.64
Add to cart
19-167-975
View Documents
KNF™ FLEXPAK
020500396
72 in. 60 x 40 in. (152.4 x 101.6cm)
Each for $110.97
Add to cart
19-167-976
View Documents
KNF™ FLEXPAK
020500398
72 in. 84 x 40 in. (213.36 x 101.6cm)
Each for $135.93
Add to cart
19-167-973
View Documents
KNF™ FLEXPAK
020500392
72 in. 104 x 68 in. (264.16 x 172.72cm)
Each for $168.58
Add to cart
19-167-977
View Documents
KNF™ FLEXPAK
020500400
76 in. 64 x 48 in. (162.56 x 121.92cm)
Each for $48.89
Add to cart
19-167-979
View Documents
KNF™ FLEXPAK
020500404
76 in. 60 x 28 in. (152.4 x 71.12cm)
Each for $104.42
Add to cart
19-167-980
View Documents
KNF™ FLEXPAK
020500406
80 in. 48 x 48 in. (121.92 x 121.92cm)
Each for $119.60
Add to cart
19-167-982
View Documents
KNF™ FLEXPAK
020500410
80 in. 160 x 60 in. (406.4 x 152.4cm)
Each for $260.00
Add to cart
19-167-986
View Documents
KNF™ FLEXPAK
020500417
80 in. 120 x 68 in. (304.8 x 172.72cm)
Each for $219.96
Add to cart
19-167-987
View Documents
KNF™ FLEXPAK
020500419
80 in. 32 x 28 in. (81.28 x 71.12cm)
Each for $87.05
Add to cart
19-167-990
View Documents
KNF™ FLEXPAK
020500427
76 in. 60 x 40 in. (152.4 x 101.6cm)
Each for $116.38
Add to cart
19-167-988
View Documents
KNF™ FLEXPAK
020500423
72 in. 128 x 48 in. (325.12 x 121.92cm)
Each for $160.99
Add to cart
19-167-999
View Documents
KNF™ FLEXPAK
020500477
72 in. 64 x 24 in. (162.56 x 60.96cm)
Each for $99.01
Add to cart
19-130-2136
View Documents
KNF™ FLEXPAK
020500489
80 in. 44 x 60 in. (111.76 x 152.4cm)
Each for $142.48
Add to cart
19-168-002
View Documents
KNF™ FLEXPAK
020500501
72 in. 64 x 40 in. (162.56 x 101.6cm)
Each for $94.64
Add to cart
19-168-005
View Documents
KNF™ FLEXPAK
020500506
76 in. 72 x 56 in. (182.88 x 142.24cm)
Each for $146.85
Add to cart
19-168-016
View Documents
KNF™ FLEXPAK
020500630
76 in. 88 x 68 in. (223.52 x 172.72cm)
Each for $122.93
Add to cart
19-168-021
View Documents
KNF™ FLEXPAK
020500665
80 in. 96 x 32 in. (243.84 x 81.28cm)
Each for $154.44
Add to cart
19-130-2167
View Documents
KNF™ FLEXPAK
020500786
72 in. 72 x 32 in. (182.88 x 81.28cm)
Each for $131.66
Add to cart
19-168-027
View Documents
KNF™ FLEXPAK
020500836
80 in. 136 x 92 in. (345.44 x 233.68cm)
Each for $205.50
Add to cart
19-130-2185
View Documents
KNF™ FLEXPAK
020500859
72 in. 72 x 56 in. (182.88 x 142.24cm)
Each for $183.77
Add to cart
19-130-2187
View Documents
KNF™ FLEXPAK
020500916
72 in. 60 x 68 in. (152.4 x 172.72cm)
Each for $187.10
Add to cart
19-130-2188
View Documents
KNF™ FLEXPAK
020500917
80 in. 44 x 28 in. (111.76 x 71.12cm)
Each for $109.82
Add to cart
19-130-2193
View Documents
KNF™ FLEXPAK
020500948
72 in. 40 x 40 in. (101.6 x 101.6cm)
Each for $103.38
Add to cart
19-168-035
View Documents
KNF™ FLEXPAK
020500959
80 in. 104 x 72 in. (264.16 x 182.88cm)
Each for $151.22
Add to cart
19-168-037
View Documents
KNF™ FLEXPAK
020500967
72 in. 40 x 28 in. (101.6 x 71.12cm)
Each for $76.96
Add to cart
19-168-038
View Documents
KNF™ FLEXPAK
020500968
72 in. 32 x 24 in. (81.28 x 60.96cm)
Each for $84.86
Add to cart
19-168-043
View Documents
KNF™ FLEXPAK
020500995
72 in. 80 x 36 in. (203.2 x 91.44cm)
Each for $101.19
Add to cart
19-168-044
View Documents
KNF™ FLEXPAK
020500996
72 in. 92 x 36 in. (233.68 x 91.44cm)
Each for $122.93
Add to cart
19-168-050
View Documents
KNF™ FLEXPAK
020501170
76 in. 80 x 44 in. (203.2 x 111.76cm)
Each for $113.15
Add to cart
19-130-2216
View Documents
KNF™ FLEXPAK
020501642
72 in. 84 x 36 in. (213.36 x 91.44cm)
Each for $151.22
Add to cart
19-130-2225
View Documents
KNF™ FLEXPAK
020501643
72 in. 84 x 80 in. (213.36 x 203.2cm)
Each for $208.50
Add to cart
19-130-2214
View Documents
KNF™ FLEXPAK
020501647
76 in. 40 x 28 in. (101.6 x 71.12cm)
Each for $104.42
Add to cart
19-130-2215
View Documents
KNF™ FLEXPAK
020501663
80 in. 60 x 28 in. (152.4 x 71.12cm)
Each for $157.66
Add to cart
19-168-061
View Documents
KNF™ FLEXPAK
020501825
78 in. 128 x 96 in. (325.12 x 243.84cm)
Each for $212.16
Add to cart
19-168-065
View Documents
KNF™ FLEXPAK
020575026
80 in. 36 x 28 in. (91.44 x 71.12cm)
Each for $72.90
Add to cart
19-168-066
View Documents
KNF™ FLEXPAK
020575038
72 in. 44 x 37 in. (111.76 x 93.98cm)
Each for $72.90
Add to cart
19-165-343
missing translation for 'viewDocuments'
KNF™ FLEXPAK
020500218B
80 in. 28 x 24 in. (71.12 x 60.96cm)
Pack of 10 for $604.76
Add to cart
19-168-060
missing translation for 'viewDocuments'
KNF™ FLEXPAK
020501824
72 in. 48 x 48 in. (121.92 x 121.92cm)
Each for $96.82
Add to cart
19-167-959
missing translation for 'viewDocuments'
KNF™ FLEXPAK
020500364
80 in. 40 x 28 in. (101.6 x 71.12cm) This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description
Description
Constructed from proven low-density polyethylene film. Perform well in a broad spectrum of applications. Ideal for parts that require a moderate level (Levels II and III) of packaging cleanliness.
  • Tough yet flexible
  • Chemically resistant to solvents
  • Exhibit low water vapor transmission rate and good electrical properties
Certifications
Cleaned and packaged in a Class 10/100 cleanroom, and certified to KNF Cleanroom Products Cleanliness Level II or III. FDA approved for use with food.
Compliance
Suitable for Class 100 cleanroom environments
Specifications
Specifications
PE (Polyethylene)