Learn More
KNF™ FLEXPAK Square-Bottom Antistatic Cleanroom Bags
Square-bottom, antistatic bags are treated with an amine-free agent
$83.82 - $686.40
Specifications
Material PE (Polyethylene)
View More Specs
Products 15
Catalog Number Mfr. No. Height (English) Dimensions (L x W) Price Quantity & Availability  
Catalog Number Mfr. No. Height (English) Dimensions (L x W) Price Quantity & Availability  
19-130-2134
View Documents
KNF™ FLEXPAK
020500366
84 in. 28 x 44 in. (71.12 x 111.76cm)
Each for $121.78
Add to cart
19-165-347
View Documents
KNF™ FLEXPAK
020500461B
84 in. 48 x 44 in. (121.92 x 111.76cm)
Pack of 5 for $410.28
Add to cart
19-168-036
View Documents
KNF™ FLEXPAK
020500960
84 in. 36 x 36 in. (91.44 x 91.44cm)
Each for $83.82
Add to cart
19-130-2217
View Documents
KNF™ FLEXPAK
020501672
88 in. 72 x 36 in. (182.88 x 91.44cm)
Each for $183.77
Add to cart
19-167-978
View Documents
KNF™ FLEXPAK
020500402
88 in. 80 x 52 in. (203.2 x 132.08cm)
Each for $151.22
Add to cart
19-130-2164
View Documents
KNF™ FLEXPAK
020500461
84 in. 44 x 48 in. (111.76 x 121.92cm)
Each for $141.44
Add to cart
19-168-015
View Documents
KNF™ FLEXPAK
020500626
84 in. 84 x 36 in. (213.36 x 91.44cm)
Each for $150.07
Add to cart
19-168-019
View Documents
KNF™ FLEXPAK
020500633
84 in. 92 x 60 in. (233.68 x 152.4cm)
Each for $135.93
Add to cart
19-130-2169
View Documents
KNF™ FLEXPAK
020500791
84 in. 40 x 44 in. (101.6 x 111.76cm)
Each for $132.70
Add to cart
19-168-026
View Documents
KNF™ FLEXPAK
020500798
84 in. 88 x 28 in. (223.52 x 71.12cm)
Each for $117.52
Add to cart
19-168-030
View Documents
KNF™ FLEXPAK
020500876
88 in. 124 x 92 in. (314.96 x 233.68cm)
Each for $206.65
Add to cart
19-130-2220
View Documents
KNF™ FLEXPAK
020501649
84 in. 92 x 32 in. (233.68 x 81.28cm)
Each for $188.14
Add to cart
19-165-346
View Documents
KNF™ FLEXPAK
020500366B
84 in. 44 x 28 in. (111.76 x 71.12cm)
Case of 10 for $686.40
Add to cart
19-165-355
View Documents
KNF™ FLEXPAK
020500960B
84 in. 36 x 36 in. (91.44 x 91.44cm)
Case of 10 for $677.56
Add to cart
19-165-360
View Documents
KNF™ FLEXPAK
020501672B
88 in. 72 x 36 in. (182.88 x 91.44cm)
Pack of 5 for $518.96
Add to cart
Description
Description
Constructed from proven low-density polyethylene film. Perform well in a broad spectrum of applications. Ideal for parts that require a moderate level (Levels II and III) of packaging cleanliness.
  • Tough yet flexible
  • Chemically resistant to solvents
  • Exhibit low water vapor transmission rate and good electrical properties
Certifications
Cleaned and packaged in a Class 10/100 cleanroom, and certified to KNF Cleanroom Products Cleanliness Level II or III. FDA approved for use with food.
Compliance
Suitable for Class 100 cleanroom environments
Specifications
Specifications
PE (Polyethylene)