LABOPORT™ Chemically Resistant Dry Vacuum Pumps: 20L/min. Flowrate Available on GSA/VA Contract for Federal Government customers only.

Quiet, reliable operation without the hazards associated with handling messy pump oils or contaminated wastewater generated by aspirators

Manufacturer: KNF NEUBERGER INC UN820FTP115V

 View more versions of this product

Catalog No. 13-878-42

  Check Availability
Add to Cart

    Specifications

    Amperage 1.9A
    Electrical Requirements 115V 60Hz
    Height (English) 8 in.
    Height (Metric) 20.3cm
    Hertz 60Hz
    Material PTFE, FFPM
    Model UN820FTP
    Width (English) Exterior 6.25 in.
    Width (English) 6.25 in.
    Height (English) Exterior 8 in.
    Length (English) Exterior 12.25 in.
    Temperature Range (Metric) 5° to 40°C
    Voltage 115V

  • Description & Specifications
  • Accessories

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit