Learn More
LABOPORT™ Chemically Resistant Dry Vacuum Pumps: 20L/min. Flowrate Available on GSA/VA Contract for Federal Government customers only.
Quiet, reliable operation without the hazards associated with handling messy pump oils or contaminated wastewater generated by aspirators
Manufacturer:  KNF NEUBERGER INC N820 FTP230
 View more versions of this product
Catalog No. 13-878-43
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
230/50Hz
PTFE, FFPM
Vacuum pump
8″
N820FTP
6.25″
Accessories