Learn More
LABOPORT™ Chemically Resistant Dry Vacuum Pumps: 34L/min. Flowrate Available on GSA/VA Contract for Federal Government customers only.
Quiet, reliable operation without the hazards associated with handling messy pump oils or contaminated wastewater generated by aspirators
Manufacturer:  KNF NEUBERGER INC UN840.3FTP230V
 View more versions of this product
Catalog No. 13-881-02
  Check Availability
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Add to cart
Specifications Description & Specifications
Specifications
8 in.
5° to 40°C
4.2A
8 in.
60Hz
UN840.3FTP
6.75 in.
13 in.
230V
230V 60Hz
20.3cm
PTFE, FFPM
6.75 in.
Accessories