Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LABOPORT™ Filtration Vacuum Pumps: 30L/min. Flowrate Available on GSA/VA Contract for Federal Government customers only.

Compact and lightweight vacuum pumps designed for most common laboratory applications

Manufacturer:  KNF NEUBERGER INC UN816.1.2KT.45P115V

 View more versions of this product

Catalog No. 13-880-33


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

115V 60Hz
0.25 in.
5.5 in.
14.25 in.
5° to 40°C
8.7 lb.
3.5 in.
60Hz
5.55 in.
4.01 in.
36.1cm
Vacuum gauge and regulator assembly
40°C
30L/min.
0.63cm
20
PPS, PTFE coated, FFPM
100w
3.95kg
115V
PPS
14.1cm
14.21 in.
0.6A
104°F
7.4psi
Accessories
Promotions

Promotions

  • Bel-Art Products
    Purchase a KNF™ LABOPORT™ Filtration Vacuum Pump and get a Bel-Art™ SP Scienceware™ 28-Piece Fittings Assortment Kit at no additional cost. This is an NLSU exclusive offer. You must apply and qualify for the program to be eligible.

    12-31-2017

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit