Learn More
KNF™ Neuberger LIQUIPORT™ NF Series Diaphragm Liquid Pumps Standard, PVDF Head Available on GSA/VA Contract for Federal Government customers only.
For convenient transfer and dosing of neutral or corrosive liquids
$1348.00 - $2220.00
Specifications
Certifications/Compliance CE, EN 61010-1, EN 61326-1
Electrical Requirements 100/230 V 50/60 Hz
Frequency 50/60 Hz
Includes Liquiport™ Laboratory pump, Mains plug, Operating manual
Enclosure Rating IP65
View More Specs
Products 3
Catalog Number Mfr. No. Model Maximum Pressure Flow Rate Price Quantity & Availability  
Catalog Number Mfr. No. Model Maximum Pressure Flow Rate Price Quantity & Availability  
13-880-41
View Documents
KNF™ Neuberger
NF100TT.18S
NF100TT.18S 15 psi 0.2 to 1.3 L/min.
Each for $1,348.00
Add to cart
13-880-47 Available on GSA/VA Contract for Federal Government customers only.
View Documents
KNF™ Neuberger
NF300TT.18S
NF300TT.18S 15 psi 0.5 to 3 L/min.
Each for $1,843.00
Add to cart
13-880-50
View Documents
KNF™ Neuberger
NF1.300TT.18S
NF1.300TT.18S 90 psi 0.5 to 3 L/min.
Each for $2,220.00
Add to cart
Description
Description
Pumps are self-priming, maintenance-free and will operate dry indefinitely without damage. Select the best pump based on the desired flowrate, maximum outlet pressure and the wetted materials.
  • Excellent corrosion resistance
  • Manual front-panel flowrate control
  • Pressure limit protection
  • No metal wetted parts to corrode
  • Reliable splash-proof housing
  • All models feature PTFE diaphragms and FFPM valves
Three considerations for any liquid diaphragm application:
  • The inlet of pump should not be lower than the level of liquid being transferred
  • An inlet filter is required if media contains particulates or fibers larger than 50 microns
  • Pulsation varies between specific pump models
Order Info
For 100/230V 50/60Hz.
Specifications
Specifications
CE, EN 61010-1, EN 61326-1
50/60 Hz
IP65
40°C
<40 dB (A)
100/230 V 50/60 Hz
Liquiport™ Laboratory pump, Mains plug, Operating manual
104°F
3000 rpm