Learn More
SIMDOS™ 10 Diaphragm Liquid Metering Pump Available on GSA/VA Contract for Federal Government customers only.
Compact, reliable, high-performance replacement for maintenance-intensive perstaltic pumps
Manufacturer:  KNF NEUBERGER INC FEM1.10FT.18S
 View more versions of this product
Catalog No. 13-880-416
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
100/240V 50/60Hz
0.6 in.
65
UFEM1.10FT.18S
5° to 40°C
2.2 lb.
Manual
50/60Hz
5.5 in.
3.42 in.
13cm
104°F
200rpm
CE, EN 61010-1, EN 61326-1
1 to 100L/min.
1.5cm
PTFE pump head, FFKM valve plate/seals, PTFE coated Diaphragm, Polypropylene housing
<40dBA
10w
1kg
100/240V
PTFE
14cm
5.1 in.
Mains plug, brief user guide, operating manual CD-ROM, connection kit hose and screw connectors
40°C
90psi