Learn More
Streamlight™ Knucklehead™ Div 2 Spot Alkaline Orange
Increase range of visibility with a 210 degree articulating head that rotates a full 360 degrees. STREAMLIGHT INC.™ Knucklehead™ Div 2 Spot Alkaline Orange is a lightweight, alkaline powered, hand-held work light that features an extremely strong magnet and more.
Manufacturer:  Streamlight™ 90744
Catalog No. 19-823-012
Add to cart
Description
Description
  • High Lumens of 180
  • Run Time on High of up to 3.5 hours
  • Run Time on Low of up to 16.00 hours
  • Beam Distance of 210 meters
  • Industrial rechargeable Ni-Cd 4.8 volt battery
  • IPX4 rated for water resistant operation
  • Comes with a clip, microprocessor with controlled high and low intensity modes, an emergency flash mode, and a moonlight mode
Specifications
Specifications
Work light
Nylon
18.3 oz.
7.2cm
Flashlight
23.4cm
Spot light
518.8g
Yellow